Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marcks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008538.2 | NP_032564.1 | 1, 2 | 501 | ||
GenBank mRNA | BC002068.1 | - | 1, 2 | 514 | ||
BC039911.1 | - | 1, 2 | 506 | |||
BC046601.1 | - | 1, 2 | 462 | 6311439 | ||
M60474.1 | - | 1, 2 | 490 | |||
X59374.1 | - | 1, 2 | 392 |