Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008539.3 | NP_032565.2 | 2 | 471 | ||
GenBank mRNA | AK017583.1 | - | 2 | 432 | ||
AK034045.1 | - | 2 | 432 | |||
AK054104.1 | - | 2 | 432 | |||
AK084505.1 | - | 2 | 539 | |||
AK087692.1 | - | 432 | ||||
AK158509.1 | - | 2 | 534 | |||
BC058693.1 | - | 2 | 471 | 6811514 | ||
U58992.1 | - | 1 | 144 | |||
U74359.1 | - | 2 | 337 |