Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008540.2 | NP_032566.2 | 3 | 713 | ||
GenBank mRNA | AB221585.1 | - | 2 | 273 | ||
AK004804.1 | - | 2 | 77 | |||
AY493561.1 | - | 2 | 273 | |||
BC046584.1 | - | 3 | 713 | 6313280 | ||
GU072917.1 | - | 2 | 273 | |||
GU072918.1 | - | 2 | 273 | |||
GU072919.1 | - | 2 | 273 | |||
GU072920.1 | - | 2 | 273 | |||
U79748.1 | - | 2 | 593 |