Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318014.1 | NP_001304943.1 | 4 | 1150 | ||
NM_001318022.1 | NP_001304951.1 | 4 | 1033 | |||
NM_030883.4 | NP_112145.1 | 4 | 1037 | |||
XM_011514470.2 | XP_011512772.1 | 5 | 2434 | |||
XM_011514472.2 | XP_011512774.1 | 4 | 1791 | |||
XM_011514473.2 | XP_011512775.1 | 5 | 1886 | |||
XM_011514479.2 | XP_011512781.1 | 3 | 1627 | |||
XM_011514483.2 | XP_011512785.1 | 3 | 1008 | |||
XM_011514484.2 | XP_011512786.1 | 2 | 1006 | |||
XM_017010737.1 | XP_016866226.1 | 4 | 2537 | |||
XM_017010738.1 | XP_016866227.1 | 4 | 1918 | |||
XM_017010739.1 | XP_016866228.1 | 4 | 2537 | |||
XM_017010740.1 | XP_016866229.1 | 5 | 2284 | |||
GenBank mRNA | AJ459845.1 | - | 2 | 564 | ||
AJ459846.1 | - | 2 | 577 | |||
AJ459847.1 | - | 1 | 542 | |||
AK127984.1 | - | 2 | 1251 | |||
BC048991.2 | - | 3 | 985 | 5271584 |