Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164041.1 | NP_001157513.1 | 2 | 768 | ||
NM_001164042.1 | NP_001157514.1 | 2 | 388 | |||
NM_008541.3 | NP_032567.1 | 2 | 365 | |||
GenBank mRNA | AF063006.1 | - | 2 | 769 | ||
AK018077.1 | - | 2 | 337 | |||
AK082997.1 | - | 2 | 389 | |||
AK139765.1 | - | 2 | 310 | |||
BC048233.1 | - | 2 | 276 | |||
BC050001.1 | - | 2 | 268 | 6490853 | ||
U58993.1 | - | 2 | 344 | |||
U77638.1 | - | 2 | 514 |