Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008548.4 | NP_032574.1 | 2 | 963 | ||
GenBank mRNA | AK028996.1 | - | 2 | 878 | ||
AK031038.1 | - | 2 | 964 | |||
AK042371.1 | - | 2 | 899 | |||
AK043932.1 | - | 2 | 904 | |||
AK148407.1 | - | 2 | 904 | |||
AK154820.1 | - | 2 | 905 | |||
AK171841.1 | - | 2 | 906 | |||
AK172376.1 | - | 2 | 905 | |||
AK172667.1 | - | 2 | 905 | |||
BC015265.1 | - | 2 | 864 | 4223319 | ||
U04299.1 | - | 1 | 567 |