Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mc1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008559.2 | NP_032585.2 | 1 | 1404 | ||
GenBank mRNA | AK148437.1 | - | 1 | 1381 | ||
BC119294.1 | - | 1 | 1078 | 8734043 | ||
BC119296.1 | - | 1 | 1078 | 8734045 | ||
X65635.1 | - | 1 | 990 |
Set Membership: |
G-protein coupled receptor |