Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008453.5 | NP_032479.1 | 5 | 1077 | ||
GenBank mRNA | AK007959.1 | - | 5 | 1076 | ||
AK081467.1 | - | 3 | 561 | |||
AK143838.1 | - | 5 | 1080 | |||
AK157576.1 | - | 5 | 1211 | |||
BC116938.1 | - | 5 | 1044 | 40126257 | ||
BC119214.1 | - | 5 | 1044 | 8733963 | ||
DQ981866.1 | - | 5 | 972 | |||
U36340.1 | - | 4 | 813 |