Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008459.2 | NP_032485.2 | 1, 2 | 128 | ||
GenBank mRNA | AF110492.1 | - | 1, 2 | 128 |