Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008465.5 | NP_032491.2 | 5 | 548 | ||
GenBank mRNA | AK028259.1 | - | 5 | 549 | ||
AK028307.1 | - | 5 | 567 | |||
AK154800.1 | - | 5 | 566 | |||
AK169311.1 | - | 5 | 571 | |||
AK169348.1 | - | 5 | 571 | |||
AK195236.1 | - | 4 | 292 | |||
BC006771.1 | - | 5 | 533 | 3588534 | ||
U20619.1 | - | 5 | 553 | |||
U34228.1 | - | 4 | 397 |