Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002257.1 | - | 6 | 722 | ||
AK019391.1 | - | 6 | 759 | |||
AK131637.1 | - | 6 | 747 | |||
BC046755.1 | - | 6 | 744 | 5693226 | ||
BC148214.1 | - | 2 | 196 | |||
CX208147.1 | - | 6 | 732 | |||
X51905.1 | - | 6 | 693 |