Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008493.3 | NP_032519.1 | 2 | 189 | ||
GenBank mRNA | AK030984.1 | - | 2 | 189 | ||
AK142589.1 | - | 2 | 192 | |||
BC125245.1 | - | 1 | 160 | 40054245 | ||
FJ374142.1 | - | 1 | 130 | |||
HQ166716.1 | - | 2 | 189 | |||
U18812.1 | - | 2 | 186 |