Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008514.4 | NP_032540.2 | 4 | 888 | ||
GenBank mRNA | AF074265.1 | - | 4 | 757 | ||
BC060704.1 | - | 4 | 888 | 6409058 | ||
CJ124877.1 | - | 3 | 546 |