Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008516.4 | NP_032542.1 | 2 | 1290 | ||
GenBank mRNA | AK139355.1 | - | 2 | 1248 | ||
AK139965.1 | - | 2 | 1292 | |||
AK160841.1 | - | 2 | 1169 | |||
AK164849.1 | - | 2 | 1249 | |||
AK173186.1 | - | 1 | 535 | |||
BC031122.1 | - | 1 | 565 | 4972803 | ||
BC058701.1 | - | 2 | 1223 | 6853964 | ||
D45913.1 | - | 2 | 1236 |