Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008534.3 | NP_032560.2 | 2 | 350 | ||
GenBank mRNA | AF244131.1 | - | 2 | 350 | ||
AK040306.1 | - | 2 | 301 | |||
AK042288.1 | - | 2 | 289 | |||
AK087345.1 | - | 2 | 270 | |||
AK134106.1 | - | 2 | 367 | |||
AK150121.1 | - | 2 | 277 | |||
BC055380.1 | - | 2 | 250 | |||
BC066212.1 | - | 2 | 238 | |||
M84412.1 | - | 2 | 214 |