Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008465.5 | NP_032491.2 | 14 | 1644 | ||
GenBank mRNA | AK028259.1 | - | 14 | 1645 | ||
AK028307.1 | - | 14 | 1663 | |||
AK154800.1 | - | 14 | 1662 | |||
AK169311.1 | - | 14 | 1667 | |||
AK169348.1 | - | 14 | 1667 | |||
AK203891.1 | - | 3 | 311 | |||
BC006771.1 | - | 14 | 1629 | 3588534 | ||
U20619.1 | - | 14 | 1649 | |||
U34228.1 | - | 13 | 1493 |