Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008466.5 | NP_032492.1 | 17 | 1671 | ||
GenBank mRNA | AF020772.1 | - | 17 | 1515 | ||
AK049373.1 | - | 17 | 1542 | |||
AK161664.1 | - | 17 | 1601 | |||
BC026885.1 | - | 17 | 1565 | 3979788 |