Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008469.2 | NP_032495.2 | 4 | 825 | ||
GenBank mRNA | BC057934.1 | - | 4 | 804 | 5251453 | |
CN456368.1 | - | 4 | 460 |