Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008493.3 | NP_032519.1 | 3 | 2406 | ||
GenBank mRNA | AK030984.1 | - | 3 | 2406 | ||
AK142589.1 | - | 3 | 2409 | |||
BC038162.1 | - | 1 | 906 | |||
HQ166716.1 | - | 3 | 2406 | |||
U18812.1 | - | 3 | 2385 |