Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008523.2 | NP_032549.2 | 9 | 1192 | ||
NM_203345.2 | NP_976220.2 | 15 | 2038 | |||
NM_206941.1 | NP_996824.1 | 10 | 1375 | |||
NM_206942.2 | NP_996825.2 | 14 | 1855 | |||
GenBank mRNA | AK157478.1 | - | 7 | 1189 | ||
BC047138.1 | - | 9 | 1192 | 6389834 | ||
BC055898.1 | - | 8 | 1056 | |||
BC141299.1 | - | 14 | 1831 | 9055911 | ||
M90470.1 | - | 15 | 2038 | |||
X07984.1 | - | 9 | 1166 | |||
X52621.1 | - | 9 | 1181 |