Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277968.1 | NP_001264897.1 | 2 | 347 | ||
NM_008534.3 | NP_032560.2 | 3 | 677 | |||
GenBank mRNA | AB196804.1 | - | 3 | 590 | ||
AB196805.1 | - | 3 | 590 | |||
AB196806.1 | - | 3 | 590 | |||
AB196807.1 | - | 3 | 590 | |||
AF244130.1 | - | 3 | 677 | |||
AF244131.1 | - | 3 | 677 | |||
AK040306.1 | - | 3 | 628 | |||
AK042288.1 | - | 3 | 616 | |||
AK087345.1 | - | 3 | 597 | |||
AK088815.1 | - | 2 | 296 | |||
AK134106.1 | - | 3 | 694 | |||
AK150121.1 | - | 3 | 604 | |||
BC055380.1 | - | 3 | 577 | |||
BC066212.1 | - | 3 | 565 | |||
BC095921.1 | - | 3 | 600 | 4935844 | ||
M84412.1 | - | 3 | 541 |