Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008539.3 | NP_032565.2 | 7 | 2031 | ||
GenBank mRNA | AK017583.1 | - | 7 | 1992 | ||
AK054104.1 | - | 7 | 1992 | |||
AK158509.1 | - | 7 | 2047 | |||
BC058693.1 | - | 7 | 2031 | 6811514 | ||
U74359.1 | - | 7 | 1898 |