Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008548.4 | NP_032574.1 | 13 | 3013 | ||
GenBank mRNA | AK031038.1 | - | 13 | 3014 | ||
AK154820.1 | - | 13 | 2955 | |||
AK171841.1 | - | 13 | 2956 |