Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008574.3 | NP_032600.2 | 1 | 181 | ||
GenBank mRNA | AK077172.1 | - | 1 | 152 | ||
BC055104.1 | - | 1 | 151 | 6773089 | ||
BC061167.1 | - | 1 | 131 | 6771333 | ||
BU936528.1 | - | 1 | 184 | |||
M29603.1 | - | 1 | 154 | |||
S49659.1 | - | 1 | 151 | |||
S49660.1 | - | 301 |