Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008580.4 | NP_032606.4 | 2, 3 | 697 | ||
GenBank mRNA | AB006787.3 | - | 2, 3 | 749 | ||
BC116627.1 | - | 2, 3 | 697 | 40086293 | ||
BC133697.1 | - | 2, 3 | 697 | 40086283 |