Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008595.2 | NP_032621.1 | 1, 2 | 423 | ||
GenBank mRNA | AF015769.1 | - | 1, 2 | 250 | ||
AK143523.1 | - | 1, 2 | 457 | |||
BC010983.1 | - | 1, 2 | 445 | 3980692 | ||
U94349.1 | - | 1, 2 | 423 |