Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008634.2 | NP_032660.2 | 2 | 354 | ||
GenBank mRNA | AK158307.1 | - | 2 | 313 | ||
AK165800.1 | - | 2 | 271 | |||
BC019183.1 | - | 2 | 286 | |||
BC037105.1 | - | 2 | 299 | |||
BC046287.1 | - | 2 | 275 | |||
BC052016.1 | - | 2 | 275 | |||
BC078457.1 | - | 2 | 280 | |||
BC138033.1 | - | 2 | 284 | 8861052 | ||
BC138034.1 | - | 2 | 284 | 8861053 | ||
BY003859.1 | - | 2 | 354 | |||
X51396.1 | - | 2 | 292 |