Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008636.4 | NP_032662.3 | 2 | 237 | ||
GenBank mRNA | AK012676.1 | - | 2 | 205 | ||
AK030284.1 | - | 2 | 216 | |||
AK150295.1 | - | 3 | 299 | |||
AK208188.1 | - | 1 | 99 | |||
AK211875.1 | - | 1 | 99 | |||
AK213643.1 | - | 1 | 99 | |||
AY260849.1 | - | 1 | 91 | |||
AY260850.1 | - | 1 | 91 | |||
BC017679.1 | - | 2 | 199 | 4210829 | ||
DV658365.1 | - | 2 | 256 | |||
X71327.1 | - | 2 | 187 |