Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008654.2 | NP_032680.1 | 2 | 420 | ||
GenBank mRNA | AK079339.1 | - | 2 | 377 | ||
AK136336.1 | - | 2 | 377 | |||
AK154697.1 | - | 1 | 902 | |||
AK167317.1 | - | 2 | 376 | |||
BC138485.1 | - | 2 | 285 | 8861505 | ||
BC138486.1 | - | 2 | 285 | 8861506 | ||
X51829.1 | - | 2 | 365 |