Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053214.2 | NP_444444.2 | 5, 6 | 563 | ||
GenBank mRNA | AK153908.1 | - | 5, 6 | 526 | ||
AK158333.1 | - | 5, 6 | 478 | |||
BC046502.1 | - | 5, 6 | 551 | 4167558 | ||
X97650.1 | - | 5, 6 | 481 |