Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008565.3 | NP_032591.3 | 6, 7 | 918 | ||
GenBank mRNA | AK036653.1 | - | 6, 7 | 878 | ||
AK083618.1 | - | 5, 6 | 917 | |||
AK088796.1 | - | 6, 7 | 838 | |||
AK089999.1 | - | 6, 7 | 822 | |||
AK135422.1 | - | 6, 7 | 820 | |||
AK141132.1 | - | 6, 7 | 884 | |||
AK141644.1 | - | 6, 7 | 919 | |||
AK146662.1 | - | 6, 7 | 841 | |||
AK151499.1 | - | 6, 7 | 832 | |||
AK157725.1 | - | 6, 7 | 820 | |||
AK220442.1 | - | 6, 7 | 820 | |||
BC013094.1 | - | 6, 7 | 874 | 3584640 | ||
D26089.1 | - | 6, 7 | 824 |