Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008568.2 | NP_032594.1 | 3, 4 | 387 | ||
GenBank mRNA | AK132748.1 | - | 948 | |||
AK133091.1 | - | 3, 4 | 383 | |||
AK144980.1 | - | 3, 4 | 399 | |||
AK154052.1 | - | 3, 4 | 384 | |||
BC023232.1 | - | 3, 4 | 363 | |||
BC065164.1 | - | 3, 4 | 371 | 6829737 | ||
BC066024.1 | - | 3, 4 | 326 | 6829337 | ||
D26091.1 | - | 3, 4 | 387 |