Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfge8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045489.1 | NP_001038954.1 | 2 | 270 | ||
NM_008594.2 | NP_032620.2 | 2 | 270 | |||
GenBank mRNA | AB021130.2 | - | 2 | 177 | ||
AB025280.1 | - | 2 | 177 | |||
AK089211.1 | - | 2 | 236 | |||
AK145183.1 | - | 2 | 234 | |||
AK148371.1 | - | 2 | 259 | |||
AK152088.1 | - | 2 | 259 | |||
AK155047.1 | - | 2 | 268 | |||
AK169997.1 | - | 2 | 234 | |||
AK170935.1 | - | 2 | 234 | |||
AK171143.1 | - | 2 | 234 | |||
AK193016.1 | - | 1 | 112 | |||
AK194326.1 | - | 1 | 112 | |||
AK195127.1 | - | 1 | 112 | |||
AK195556.1 | - | 1 | 112 | |||
AK197795.1 | - | 1 | 112 | |||
AK200279.1 | - | 1 | 112 | |||
AK200283.1 | - | 1 | 112 | |||
AK200926.1 | - | 1 | 112 | |||
AK202028.1 | - | 1 | 112 | |||
AK202981.1 | - | 1 | 112 | |||
AK203389.1 | - | 1 | 112 | |||
AK205167.1 | - | 1 | 112 | |||
AK210333.1 | - | 1 | 112 | |||
BC003892.1 | - | 2 | 214 | 3593310 | ||
BC003904.1 | - | 2 | 209 | 3601565 | ||
BC018577.1 | - | 2 | 207 | |||
M38337.1 | - | 2 | 222 | |||
Y11684.2 | - | 2 | 177 |