Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008637.1 | NP_032663.1 | 3 | 230 | ||
GenBank mRNA | AK011695.1 | - | 3 | 219 | ||
AK088309.1 | - | 3 | 222 | |||
AK168312.1 | - | 3 | 233 | |||
BC021940.1 | - | 2 | 201 | 5067377 | ||
BC098239.1 | - | 2 | 200 | 3469785 | ||
D49956.1 | - | 3 | 230 |