Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008565.3 | NP_032591.3 | 12, 13 | 2153 | ||
GenBank mRNA | AK011743.1 | - | 12, 13 | 2088 | ||
AK036653.1 | - | 12, 13 | 2113 | |||
AK088796.1 | - | 12, 13 | 2073 | |||
AK089999.1 | - | 12, 13 | 2057 | |||
AK135422.1 | - | 12, 13 | 2055 | |||
AK141132.1 | - | 12, 13 | 2121 | |||
AK141644.1 | - | 12, 13 | 2154 | |||
AK146662.1 | - | 12, 13 | 2076 | |||
AK151499.1 | - | 12, 13 | 2067 | |||
AK157725.1 | - | 12, 13 | 2055 | |||
AK220442.1 | - | 12, 13 | 2055 | |||
BC013094.1 | - | 12, 13 | 2109 | 3584640 | ||
D26089.1 | - | 12, 13 | 2059 |