Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008568.2 | NP_032594.1 | 4 | 402 | ||
GenBank mRNA | AK132748.1 | - | 963 | |||
AK133091.1 | - | 4 | 398 | |||
AK149913.1 | - | 4 | 399 | |||
AK150059.1 | - | 4 | 399 | |||
AK150066.1 | - | 4 | 399 | |||
AK154052.1 | - | 4 | 399 | |||
BC023232.1 | - | 4 | 378 | |||
BC065164.1 | - | 4 | 386 | 6829737 | ||
BC066024.1 | - | 4 | 341 | 6829337 | ||
D26091.1 | - | 4 | 402 |