Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mme | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289462.1 | NP_001276391.1 | 23 | 4403 | ||
NM_001289463.1 | NP_001276392.1 | 23 | 4187 | |||
NM_008604.4 | NP_032630.2 | 23 | 4037 | |||
GenBank mRNA | AK031446.1 | - | 23 | 4038 | ||
AK077940.1 | - | 1 | 1253 |