Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088476.1 | - | 10 | 2482 | ||
AK155117.1 | - | 10 | 2484 | |||
U54984.1 | - | 10 | 2464 | |||
X83536.2 | - | 10 | 2392 |