Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpv17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294322.1 | NP_001281251.1 | 7 | 711 | ||
NM_001294324.1 | NP_001281253.1 | 8 | 773 | |||
NM_008622.6 | NP_032648.1 | 8 | 744 | |||
GenBank mRNA | BC013452.1 | - | 7 | 697 | 4237284 | |
CF731561.1 | - | 1 | 138 | |||
M36411.1 | - | 8 | 726 |