Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008634.2 | NP_032660.2 | 5 | 2265 | ||
GenBank mRNA | AK038690.1 | - | 1 | 1086 | ||
BC019183.1 | - | 5 | 2197 | |||
BC138033.1 | - | 5 | 2195 | 8861052 | ||
BC138034.1 | - | 5 | 2195 | 8861053 | ||
X51396.1 | - | 5 | 2203 |