Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtnr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008639.2 | NP_032665.1 | 2 | 1055 | ||
GenBank mRNA | BC116900.1 | - | 2 | 1128 | 40126219 | |
BC119313.1 | - | 2 | 1128 | 8734062 | ||
U52222.1 | - | 2 | 1055 |
Set Membership: |
G-protein coupled receptor |