Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042743.2 | NP_001036208.1 | 18 | 2308 | ||
NM_008641.3 | NP_032667.2 | 17 | 2287 | |||
GenBank mRNA | AB093264.1 | - | 8 | 940 | ||
BC060703.1 | - | 18 | 1878 | 6412811 | ||
CK633885.1 | - | 3 | 336 | |||
U02313.1 | - | 17 | 1957 |