Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008652.2 | NP_032678.1 | 4, 5 | 469 | ||
GenBank mRNA | AK028497.1 | - | 4, 5 | 469 | ||
AK183670.1 | - | 1, 2 | 62 | |||
BC050842.1 | - | 4, 5 | 415 | 6518298 | ||
X70472.1 | - | 4, 5 | 417 |