Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008653.2 | NP_032679.2 | 35 | 4054 | ||
GenBank mRNA | AK146661.1 | - | 35 | 4055 | ||
AK146886.1 | - | 35 | 4055 | |||
AK169302.1 | - | 34 | 4032 | |||
AK169306.1 | - | 34 | 4032 | |||
BC094945.1 | - | 35 | 4051 |