Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053214.2 | NP_444444.2 | 19 | 2174 | ||
GenBank mRNA | AK036840.1 | - | 9 | 2007 | ||
AK131188.1 | - | 9 | 867 | |||
AK153908.1 | - | 19 | 2137 | |||
AK158333.1 | - | 19 | 2089 | |||
BC046502.1 | - | 19 | 2162 | 4167558 | ||
X97650.1 | - | 19 | 2092 |