Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008669.4 | NP_032695.3 | 8, 9 | 1091 | ||
GenBank mRNA | AF079458.1 | - | 8, 9 | 1049 | ||
AK077116.1 | - | 8, 9 | 1063 | |||
AK153002.1 | - | 8, 9 | 1077 | |||
BC021631.1 | - | 8, 9 | 1071 | 4019197 |