Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKNOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022062.2 | NP_071345.2 | 7 | 730 | ||
XM_005271642.2 | XP_005271699.1 | 6 | 619 | |||
XM_005271643.2 | XP_005271700.1 | 4 | 436 | |||
XM_006718894.2 | XP_006718957.1 | 6 | 506 | |||
XM_011542944.2 | XP_011541246.1 | 7 | 815 | |||
XM_011542945.2 | XP_011541247.1 | 8 | 775 | |||
XM_011542946.1 | XP_011541248.1 | 7 | 698 | |||
XM_011542947.2 | XP_011541249.1 | 5 | 384 | |||
XM_017018110.1 | XP_016873599.1 | 7 | 3565 | |||
XM_017018111.1 | XP_016873600.1 | 5 | 425 | |||
XM_017018112.1 | XP_016873601.1 | 4 | 436 | |||
GenBank mRNA | AB065001.1 | - | 4 | 408 | ||
AJ417081.1 | - | 4 | 380 | |||
AK023136.1 | - | 4 | 408 | |||
AK298999.1 | - | 6 | 499 | |||
AK316001.1 | - | 6 | 530 | |||
AK316268.1 | - | 5 | 518 | |||
AL512682.1 | - | 2 | 53 | |||
BC045626.1 | - | 7 | 687 | 5261106 | ||
BX648615.1 | - | 5 | 507 |