Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081453.1 | NP_001074922.1 | 1 | 71 | ||
NM_001286079.2 | NP_001273008.1 | 2 | 253 | |||
NM_001286080.2 | NP_001273009.1 | 3 | 300 | |||
NM_008697.4 | NP_032723.2 | 3 | 300 | |||
GenBank mRNA | AK129394.1 | - | 3 | 190 | ||
AK134860.1 | - | 2 | 266 | |||
AK139959.1 | - | 2 | 279 | |||
AK141891.1 | - | 2 | 241 | |||
AK162150.1 | - | 3 | 244 | |||
AK167264.1 | - | 2 | 246 | |||
AY515727.1 | - | 1 | 50 | |||
BC137789.1 | - | 2 | 138 | 8860808 | ||
BC144913.1 | - | 3 | 185 | 9053461 | ||
BC144914.1 | - | 3 | 185 | |||
CJ089455.1 | - | 2 | 246 | |||
U40342.1 | - | 3 | 216 |