Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx2-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008700.2 | NP_032726.1 | 2 | 569 | ||
GenBank mRNA | AK142146.1 | - | 2 | 474 | ||
BC139299.1 | - | 2 | 415 | 8862321 | ||
BC139303.1 | - | 2 | 415 | 8862325 | ||
L20300.1 | - | 2 | 386 | |||
X75415.1 | - | 2 | 634 |