Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008746.5 | NP_032772.3 | 1 | 238 | ||
NM_182809.2 | NP_877961.1 | 1 | 238 | |||
GenBank mRNA | AF035399.1 | - | 2 | 333 | ||
AF035400.1 | - | 1 | 387 | |||
AY336094.1 | - | 1 | 183 | |||
BC139764.1 | - | 2 | 250 | 40110345 | ||
JF417977.1 | - | 1 | 17 |